Learn More
ATCC Corynebacterium urealyticum
Supplier:  ATCC 43044
Encompass
Corynebacterium urealyticum strain 3 [Garcia strain] was isolated in Spain from urine. This whole-genome sequenced bacterium can be used as a quality control strain for the bioMerieux VITEK 2 CBC Test Kit.
Catalog No. 50-238-2981