Learn More
ATCC Corynebacterium xerosis
Catalog No. 502383553
Encompass
Corynebacterium xerosis was isolated from the ear discharge of a child. This bacterial type strain produces L-aspartyl-L-phenylalanine esters and carnitine L-carnitine.
Catalog No. 50-238-3553 Supplier ATCC Supplier No. 373
Corynebacterium xerosis was isolated from the ear discharge of a child. This bacterial type strain produces L-aspartyl-L-phenylalanine esters and carnitine L-carnitine.