Learn More
Medchemexpress LLC Corynoline | 18797-79-0 | 98.8% | 367.40 g·mol⁻¹ | C21H21NO5 | 10 MG
SDP
Catalog No. 5000295897
Encompass_Preferred
Corynoline is a natural isoquinoline alkaloid used as a research reagent. It functions as a reversible, noncompetitive acetylcholinesterase (AChE) inhibitor (IC50 = 30.6 μM) and has reported anti-inflammatory activity through activation of the Nrf2 pathway. The compound is supplied as a high-purity solid for laboratory studies and is intended for research use only.
  • Reversible, noncompetitive acetylcholinesterase inhibitor (IC50 = 30.6 μM).
  • Activates Nrf2 and exhibits anti-inflammatory activity.
  • High purity suitable for analytical and biological research.
  • Supplied as a 10 mg solid for laboratory use.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-95897 Supplier Medchemexpress LLC Supplier No. HYN082610MG
Corynoline is a natural isoquinoline alkaloid used as a research reagent. It functions as a reversible, noncompetitive acetylcholinesterase (AChE) inhibitor (IC50 = 30.6 μM) and has reported anti-inflammatory activity through activation of the Nrf2 pathway. The compound is supplied as a high-purity solid for laboratory studies and is intended for research use only.
  • Reversible, noncompetitive acetylcholinesterase inhibitor (IC50 = 30.6 μM).
  • Activates Nrf2 and exhibits anti-inflammatory activity.
  • High purity suitable for analytical and biological research.
  • Supplied as a 10 mg solid for laboratory use.
  • Intended for research use only; not for human or veterinary use.