Learn More
Medchemexpress LLC HY-N0590 5mg Medchemexpress, Corynoxeine CAS:630-94-4 Purity:>98%
SDP
Catalog No. 501933625
Encompass_Preferred
Medchemexpress, HY-N0590 5mg Corynoxeine CAS:630-94-4 Corynoxeine, isolated from the hook of Uncaria rhynchophylla, is a potent ERK1 / ERK2 inhibitor of key PDGF-BB-induced vascular smooth muscle cells (VSMCs) proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3625 Supplier Medchemexpress LLC Supplier No. HYN05905MG
Medchemexpress, HY-N0590 5mg Corynoxeine CAS:630-94-4 Corynoxeine, isolated from the hook of Uncaria rhynchophylla, is a potent ERK1 / ERK2 inhibitor of key PDGF-BB-induced vascular smooth muscle cells (VSMCs) proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.