Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Corynoxidine | 57906-85-1 | 5 MG
SDP

Catalog No. 5000418554
Encompass_Preferred

(-)-Corynoxidine is an acetylcholinesterase inhibitor isolated from the aerial parts of Corydalis speciosa. It exhibits antibacterial activities against Staphylococcus aureus and methicillin-resistant S. aureus strains. With a molecular weight of 371.43 and a formula of C21H25NO5, it appears as an off-white to light yellow solid. It is primarily for research use only.

  • Acetylcholinesterase inhibitor (IC50: 89.0 μM)
  • Isolated from Corydalis speciosa
  • Exhibits antibacterial activities
  • Molecular weight: 371.43
  • Formula: C21H25NO5
  • Appearance: Off-white to light yellow solid
  • Purity: 99.90%
  • Classified as alkaloid and isoquinoline alkaloid

Catalog No. 50-004-18554 Supplier Medchemexpress LLC Supplier No. HYN70105MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Corynoxidine is an acetylcholinesterase inhibitor isolated from the aerial parts of Corydalis speciosa. It exhibits antibacterial activities against Staphylococcus aureus and methicillin-resistant S. aureus strains. With a molecular weight of 371.43 and a formula of C21H25NO5, it appears as an off-white to light yellow solid. It is primarily for research use only.

  • Acetylcholinesterase inhibitor (IC50: 89.0 μM)
  • Isolated from Corydalis speciosa
  • Exhibits antibacterial activities
  • Molecular weight: 371.43
  • Formula: C21H25NO5
  • Appearance: Off-white to light yellow solid
  • Purity: 99.90%
  • Classified as alkaloid and isoquinoline alkaloid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.