Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Corypalmine | 27313-86-6 | 99.5% | 341.40 g/mol | C20H23NO4 | 10 MG
SDP

Catalog No. 5000295522
Encompass_Preferred

Corypalmine is a naturally occurring isoquinoline alkaloid isolated from Stephania cepharantha. It is reported to exhibit antifungal and antimicrobial activity and is supplied as a high-purity research chemical for biochemical and pharmacological studies.

  • Isoquinoline alkaloid chemical class.
  • Reported antifungal activity.
  • Molecular formula C20H23NO4.
  • Molecular weight 341.40 g/mol.
  • High purity (99.48%).
  • Supplied as a solid powder suitable for laboratory research.
  • For research use only; not for human use.

Catalog No. 50-002-95522 Supplier Medchemexpress LLC Supplier No. HYN065410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corypalmine is a naturally occurring isoquinoline alkaloid isolated from Stephania cepharantha. It is reported to exhibit antifungal and antimicrobial activity and is supplied as a high-purity research chemical for biochemical and pharmacological studies.

  • Isoquinoline alkaloid chemical class.
  • Reported antifungal activity.
  • Molecular formula C20H23NO4.
  • Molecular weight 341.40 g/mol.
  • High purity (99.48%).
  • Supplied as a solid powder suitable for laboratory research.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.