Learn More
Cosmo Bio Usa Inc COS banker Cell Freezing Medium, 120 mL, serum-free, animal product-free, chemically defined, contains 10% DMSO, -80 °C storage, long-term cell storage, iPS cell freezing
SDP
Catalog No. NC2022268
Encompass
COS banker Cell Freezing Medium, 120 mL, serum-free, animal product-free, chemically defined, contains 10% DMSO, -80 °C storage, long-term cell storage, iPS cell freezing
Catalog No. NC2022268 Supplier Cosmo Bio Usa Inc Supplier No. KOJCOSCFM01
COS banker Cell Freezing Medium, 120 mL, serum-free, animal product-free, chemically defined, contains 10% DMSO, -80 °C storage, long-term cell storage, iPS cell freezing