Learn More
Ecolab COSA Oxonia Active Antimicrobial Agent, EPA Registered, Non-Foaming, 50 US gal
Catalog No. NC3502451
Encompass_Preferred
COSA™ Oxonia Active is a 50 gal EPA-registered peroxyacetic acid antimicrobial agent. It effectively sanitizes and sterilizes aseptic processing and packaging equipment, demonstrating high efficacy against various pathogens while being non-corrosive to metals. Its non-foaming formula minimizes Clean-in-Place cycle times, enhancing operational efficiency. The product is supported by a comprehensive validation package, setting it apart with features ideal for scientific and industrial environments.
Catalog No. NC3502451 Supplier Ecolab Supplier No. 6600027
COSA™ Oxonia Active is a 50 gal EPA-registered peroxyacetic acid antimicrobial agent. It effectively sanitizes and sterilizes aseptic processing and packaging equipment, demonstrating high efficacy against various pathogens while being non-corrosive to metals. Its non-foaming formula minimizes Clean-in-Place cycle times, enhancing operational efficiency. The product is supported by a comprehensive validation package, setting it apart with features ideal for scientific and industrial environments.