Learn More
Beckman Coulter CosMCPrep Plasmid Purification Kit - 384 Preps
Catalog No. NC9947315
Encompass
A SPRI paramagnetic bead-based plasmid purification system that uses a single protocol to purify a variety of high and low copy number template types. This flexible system is easily automated without sacrificing product quality. The single protocol, ease of automation and high quality results make CosMCPrep an ideal solution for all your plasmid purification needs.
Catalog No. NC9947315 Supplier Beckman Coulter Supplier No. A37064
A SPRI paramagnetic bead-based plasmid purification system that uses a single protocol to purify a variety of high and low copy number template types. This flexible system is easily automated without sacrificing product quality. The single protocol, ease of automation and high quality results make CosMCPrep an ideal solution for all your plasmid purification needs.