Learn More
PCE AMERICAS INC Portable Comestic pH Meter 0.00 to 14.00 pH
SDP
Catalog No. NC3654419
Encompass_Preferred
"The Cosmetics pH Meter PCE-228P can be used in all stages of cosmetic production. pH measurement in cosmetics is a priority test that should be performed not only to ensure that the pH of the product matches the pH of the skin, but also to ensure good quality, suitable viscosity, colour / odour and to ensure durability of the cosmetics. Measuring range: 0.00 … 14.00 pHResolution: 0.01 pHAccuracy: ± (0.02 pH + 2d)Memory: flexible via SD card memory 1 … 16 GBInterface: RS -232Applications: Shampoo, pastes, paints, cosmetics"
Catalog No. NC3654419 Supplier PCE AMERICAS INC Supplier No. PCE228P
"The Cosmetics pH Meter PCE-228P can be used in all stages of cosmetic production. pH measurement in cosmetics is a priority test that should be performed not only to ensure that the pH of the product matches the pH of the skin, but also to ensure good quality, suitable viscosity, colour / odour and to ensure durability of the cosmetics. Measuring range: 0.00 … 14.00 pHResolution: 0.01 pHAccuracy: ± (0.02 pH + 2d)Memory: flexible via SD card memory 1 … 16 GBInterface: RS -232Applications: Shampoo, pastes, paints, cosmetics"