Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 150 x 4.6 mm

Catalog No. 502396054 Shop All Phenomenex Products
Encompass

Cosmosil™ 5 m C18-AR-II CH0-4163

Catalog No. 50-239-6054 Supplier Phenomenex Inc Supplier No. CH04163
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cosmosil™ 5 m C18-AR-II CH0-4163

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.