Learn More
Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates
GSA_VAGREENER_CHOICE
Catalog No. 07200568 Shop All Corning Products
Change view
Click to view available options
Color:
White, Clear Bottom
Catalog No. Color
07-200-568 White, Clear Bottom
1 options
Catalog No. 07-200-568 Supplier Corning™ Supplier No. 3632
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates permit direct microscopic viewing. For fluorescent and luminescent assays; ideal for performing cell-based and molecular assays.
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color White, Clear Bottom
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Untreated
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov