Learn More
Description
- Minimal well-to-well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Recommended working volumes of 75 to 200μL
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 320μL to 360μL
- Working volume: 75μL to 200μL
- Choice of flat, round or V-shaped bottom
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
- Plate dimensions meet proposed industry standards
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Lid | Without Lid |
| Array | 8 x 12 |
| Binding Type | Medium Binding |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 75 to 200 μL |
| Sterility | Non-sterile |
| Surface Treatment | Untreated |
| Well Shape | Round Bottom (U-well) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

