Learn More
Corning™ 96-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE
Catalog No. 064432 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
06-443-2 Black
1 options
Catalog No. 06-443-2 Supplier Corning™ Supplier No. 3792
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Packaging Inner packs of 25
    Color Black
    Product Type EIA/RIA
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Untreated
    Well Shape Round
    For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov