Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates

Catalog No. 07200613 Shop All Corning Products
Change view
Click to view available options
Color:
Natural
Barcode:
No Barcode
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Barcode
07-200-613 Natural No Barcode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-200-613 Supplier Corning™ Supplier No. 6551
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constructed of medical grade polypropylene to accommodate wide temperature changes that might occur from storage to thermal cycling. Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.

Corning Thermowell GOLD 96-well PCR microplates are offered in four formats to ensure maximum flexibility and a perfect match for your applications. The original Thermowell 96-well PCR microplates are universal fit and can be cut into 3 x 8 well segments.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certified DNase and RNase-free
Format Standard
Lid Without Lid
Description Can be cut into 3 x 8 well strips for lower throughput applications. Available PCR plate adapter for use with ABI PRISM™ 3700 DNA Analyzer
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 200 μL
Well Shape Conical Bottom (V-well)
For Use With (Application) PCR
No. per Case 25/Cs.
No. per Pack 25/Pk.
Quantity 25/Pk.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.