Learn More
Bio Rad Laboratories COSTAR 96-WELL FLAT-BOTTOM EIA
Catalog No. 509011636
Encompass
Plate; Bio-Rad; Costar; For EIA; 96-Well; Flat bottom; Polystyrene; 5 per package; Box of 100
Catalog No. 50-901-1636 Supplier Bio Rad Laboratories Supplier No. 2240096
Plate; Bio-Rad; Costar; For EIA; 96-Well; Flat bottom; Polystyrene; 5 per package; Box of 100