Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 96-Well Clear PVC Assay Microplates
GSA_VA

Catalog No. p-152550 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
07-200-99 Round
07-200-33 Flat Bottom (F-well)
07-200-100 Conical Bottom (V-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 07-200-99 Supplier Corning™ Supplier No. 2797
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Manufactured from clear flexible Polyvinyl Chloride (PVC) for economy and visibility. Corning™ 96-Well Clear PVC Assay Microplates present an excellent solution for laboratory miniaturization. Due to their flexible nature, these microplates are not compatible with automation.

  • Not treated PVC microplates are economical microplates for solution-based assays, serial dilutions, and general storage applications.
  • Well volume of 250 μL (260 μL for V-bottom); working well volume of 50 μL to 150 μL
  • Lids are not available.
  • Order Info

    Lids are not available. Supplied in packs of 25.

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color Clear
    Material PVC
    No. of Wells 96
    Volume (Metric) Working 50 to 150 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Well Shape Round
    For Use With (Application) Solution-based assays, serial dilutions and general storage applications
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.