Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Catalog No. 07200329 Shop All Corning Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-200-329 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-329 Supplier Corning™ Supplier No. 3694
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

Color:Black; Sterility:Non-sterile; Lid:Without Lid

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode Alphanumeric
    Format Half-Area
    Lid Without Lid
    Array 8 x 12
    Color Black
    For Use With (Equipment) Plate readers
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Suitable for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.