Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 96-Well, High Binding, Flat-Bottom, Half-Area Microplate
GSA_VAGREENER_CHOICE

Catalog No. 0720037 Shop All Corning Products
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07-200-37 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-37 Supplier Corning™ Supplier No. 3690
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Color:Clear; Sterility:Non-sterile; Lid:Without Lid

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Format Half-Area
Lid Without Lid
Array 8 x 12
Binding Type High Bind
Color Clear
For Use With (Equipment) Plate readers
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 125 μL
Sterility Non-sterile
Surface Treatment High Binding
Well Shape Flat Bottom (F-well)
For Use With (Application) Ideal for immunoassays
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 100/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.