Learn More
Corning™ 8-Channel Adapter for Vacuum Aspirator
GSA_VA
Catalog No. 07200710 Shop All Corning Products
Change view
Click to view available options
Number of Channels:
8
Catalog No. Number of Channels
07-200-710 8
1 options
This item is not returnable. View return policy
Catalog No. 07-200-710 Supplier Corning™ Supplier No. 4931
  • $552.00
    $364.00 / Case of 1
    Save $188.00
    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with Corning Vacuum Aspirator. This 8-Channel Adapter can be used to remove media and solutions from 8 wells simultaneously on 96 well plate.
This is an 8-channel adapter that fits on the Aspirator (No. 4930) so that it can be used to remove media and solutions from 8 wells simultaneously on a 96 well plate. 
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Standard 1-200μL disposable pipet tips
Autoclavable Not Autoclavable
Number of Channels 8
Type Adapter, 8 channel
For Use With (Equipment) Corning Vacuum Aspirator (4930)