Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 96-Well, Cell Culture-Treated, V-Shaped-Bottom Microplate
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. 0720096 Shop All Corning Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07-200-96 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-200-96 Supplier Corning™ Supplier No. 3894
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Color:Clear; Sterility:Sterile; Lid:With Lid

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid With Lid
Culture Area 0.38 cm2
Array 8 x 12
Binding Type TC-Treated
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 0.38 mL
Well Shape Conical Bottom (V-well)
For Use With (Application) Attachment and growth of anchorage-dependent cells
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Diameter (Metric) Well 1.6 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.