Learn More
Corning™ 96-Well Clear Ultra Low Attachment Microplates
BIOPHARMAGREENER_CHOICE
Catalog No. 07201680 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-201-680 Clear
1 options
Catalog No. 07-201-680 Supplier Corning™ Supplier No. 7007
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Covalently bonded hydrogel minimizes cell attachment, protein absorption, enzyme activation and cellular activation. Corning™ 96-Well Clear Ultra Low Attachment Microplates allow for lentivirus transduction of human osteoclast precursor cells.
  • Ultra low attachment surface with covalently bonded hydrogel that minimizes cell attachment, protein absorption, enzyme activation and cellular activation
  • The surface is noncytotoxic, biologically inert and nondegradable
  • Round bottoms with 330μL total volume
  • Recommended working volumes of 75 to 200μL
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
  • Individually wrapped
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Binding Type Ultra-Low Attachment
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Ultra-low Binding
Well Shape Round
Includes Lids
Quantity 24/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov