Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ HTS Transwell™ Clear Untreated Reservoirs
GREENER_CHOICE

Catalog No. 07200685 Shop All Corning Products
Change view
Click to view available options
No. per Pack:
12/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
07-200-685 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-685 Supplier Corning™ Supplier No. 3395
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured from clear plastic for maximum visibility. Corning™ HTS Transwell™ Clear Untreated Reservoir is for use with HTS Transwell Permeable Supports.

  • For use as reservoir tray with HTS Transwell Permeable Supports
  • Not Treated for cell attachment
  • Same footprint as standard 24 well plate but with single rectangular well
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polystyrene
Sterility Sterile
Surface Treatment Non-treated
Type Cell Culture Insert System
For Use With (Equipment) HTS Transwell™ Permeable Supports
No. per Case 48/Cs.
No. per Pack 12/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.