Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ HTS Transwell™ -96 Well Permeable Support System
GREENER_CHOICE

Catalog No. p-4922169 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
No. per Pack:
1/Pk.
4/Pk.
5/Pk.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size No. per Pack
07-200-741 0.4 μm 5/Pk.
07-200-278 0.4 μm 1/Pk.
07-200-742 1 μm 5/Pk.
07-202-539 1 μm 1/Pk.
09-761-80 3 μm 1/Pk.
09-761-81 3 μm 4/Pk.
09-761-82 5 μm 4/Pk.
09-761-83 5 μm 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 07-200-741 Supplier Corning™ Supplier No. 3391
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boasts membrane area for cell growth or migration of 0.143cm2. Corning™ HTS Transwell™ 96-Well Permeable Support System is a microplate support system for an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.

  • The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
  • Choice of either polyester (PET) membrane (0.4 μm, 1.0 μm pore sizes) or polycarbonate (PC) membrane (0.4 μm, 3.0 μm, and 5.0 μm pore sizes)
  • 0.143 cm2 membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
  • Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access.
  • Optimized for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
  • The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabilizer to reduce the risk of spills during handling
  • The receiver plate isolates each well to enable 96 individual assays.
  • Sterile
  • The HTS Transwell-96 systems (0.4 μm PC, 0.4 μm PET, and 1.0 μm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid.
  • The HTS Transwell-96 plates (3.0 and 5.0 μm PC, 8.0 μm PET) are packaged with the 96-well insert plate in the 96-well receiver plate with lid. Reservoir plates may be purchased separately.
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Culture Area 0.143 cm2
Membrane TC-treated polycarbonate
Array 8 x 12
Pore Size 0.4 μm
Color Clear
Material Polycarbonate
No. of Wells 96
Volume (Metric) Working 0.23 mL
Sterility Sterile
Well Shape Flat Bottom (F-well)
Includes HTS Transwell™ unit (1 integral tray with 96 wells), reservoir plate (for cell growth steps), receiver plate (96 wells for growth or assay steps) and 2 sterile lids
No. per Case 5/Cs.
No. per Pack 5/Pk.
Quantity 5/Cs.
Diameter (Metric) Membrane 4.26 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.