Learn More
Corning™ Costar™ HTS Transwell™ -96 Well Receiver Plates
GREENER_CHOICE
Catalog No. 08757230 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
08-757-230 Black
1 options
This item is not returnable. View return policy
Catalog No. 08-757-230 Supplier Corning™ Supplier No. 3583
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accessory for the HTS Transwell™-96 Permeable Support system
  • The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
  • Choice of either polyester (PET) membrane (0.4 μm, 1.0 μm, and 8.0 μm pore sizes) or polycarbonate (PC) membrane (0.4 μm, 3.0 μm, and 5.0 μm pore sizes)
  • 0.143 cm2 membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
  • Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access.
  • Optimized for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
  • The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabilizer to reduce the risk of spills during handling
  • The receiver plate isolates each well to enable 96 individual assays.
  • Sterile
  • The HTS Transwell-96 systems (0.4 μm PC, 0.4 μm PET, and 1.0 μm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid.
  • The HTS Transwell-96 plates (3.0 and 5.0 μm PC, 8.0 μm PET) are packaged with the 96-well insert plate in the 96-well receiver plate with lid. Reservoir plates may be purchased separately.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI, SBS 1-2004 through ANSI, SBS 4-2004
Format Standard
Lid With Lid
Array 8 x 12
Color Black
No. of Wells 96
Sterility Sterile
Well Shape Flat Bottom (F-well)
No. per Case 10/Cs.
No. per Pack 10/Pk.
Quantity 10/Pk.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov