Learn More
Corning™ Costar™ Netwell™ Inserts
GSA_VAGREENER_CHOICE
Catalog No. 07200211 Shop All Corning Products
Change view
Click to view available options
For Use With (Application):
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
Catalog No. For Use With (Application)
07-200-211 Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
1 options
This item is not returnable. View return policy
Catalog No. 07-200-211 Supplier Corning™ Supplier No. 3477
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers.
Netwell Inserts have polyester (PET) mesh bottoms attached to a polystyrene ring or housing. They are used as tissue carriers, supports and strainers for culture of small organs, tissue slices, or explants at the air-media interface. They can be used to coarse filter tissue homogenates, cell suspensions, or microcarriers. Accessories allow them to be used as a handy carrier for immunocytochemical staining of tissue culture slices.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
Material Polystyrene