Learn More
Corning™ Costar™ Thermowell™ Plate Lid
Catalog No. 07200250 Shop All Corning Products
Change view
Click to view available options
Plate Compatibility:
Thermowell Polycarbonate Plates
Catalog No. Plate Compatibility
07-200-250 Thermowell Polycarbonate Plates
1 options
This item is not returnable. View return policy
Catalog No. 07-200-250 Supplier Corning™ Supplier No. 6520
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fits styles C, M, and H.
  • Polycarbonate lids are thermally stable up to 135°C and can be used on Corning™ Thermowell™ 96 well polycarbonate plates (fits Cat. No. 6511) with oil overlays
  • Lids are intended to be covers only and do not prevent evaporation
TRUSTED_SUSTAINABILITY
Specifications
Material Polycarbonate
Sterility Non-sterile
No. of Wells 96
No. per Case 25/Cs.
No. per Pack 1/Pk.
Product Type Plate Lid
Plate Compatibility Thermowell Polycarbonate Plates
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov