Learn More
Sigma Aldrich Fine Chemicals Biosciences CostarR vacuum aspirator 81EA
Catalog No. 502905077
Encompass
This is an 8-channel adapter that fits on to the Costar(R) Aspirator (CLS4930) so that it can be used to remove media and SOLUTIONons from 8 wells simultaneously on a 96 well plate.Please note Only the 8-channel adapter is supplied for product CLS4931 despite the image on this web page showing both the adapter and the Costar(R) Aspirator (CLS4930) together they are sold separately.
Catalog No. 50-290-5077 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS49311EA
This is an 8-channel adapter that fits on to the Costar(R) Aspirator (CLS4930) so that it can be used to remove media and SOLUTIONons from 8 wells simultaneously on a 96 well plate.Please note Only the 8-channel adapter is supplied for product CLS4931 despite the image on this web page showing both the adapter and the Costar(R) Aspirator (CLS4930) together they are sold separately.