Learn More
Sigma Aldrich Fine Chemicals Biosciences CostarR vacuum aspirator h1EA
Catalog No. 502905850
Encompass
Hand-held vacuum aspiration device with a wide variety of accessories available for removal of supernatants. The device attaches to your laboratory vacuum supply via a tubulation fitting. fingertip control of vacuum reduces the risk of losing valuable samples.Use with pipette tips Pasteur pipettes or any of the aspirator accessories below. Unit comes with single-channel adapter for disposable pipette tips.
Catalog No. 50-290-5850 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS49301EA
Hand-held vacuum aspiration device with a wide variety of accessories available for removal of supernatants. The device attaches to your laboratory vacuum supply via a tubulation fitting. fingertip control of vacuum reduces the risk of losing valuable samples.Use with pipette tips Pasteur pipettes or any of the aspirator accessories below. Unit comes with single-channel adapter for disposable pipette tips.