Learn More
World Precision Instrument Costotome-Rib Cutting Knife, 8in
SDP
Catalog No. 502538365
Encompass
The Costotome Rib Cutting Knife is designed slicing through cartilage in the chest cavity during gross dissection and post-mortem exams. This knife is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.
Catalog No. 50-253-8365 Supplier World Precision Instrument Supplier No. WP0189
The Costotome Rib Cutting Knife is designed slicing through cartilage in the chest cavity during gross dissection and post-mortem exams. This knife is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.