Learn More
Cayman Chemical CosyntropInacetate 10mg
SDP
Catalog No. 502994393
Encompass_Preferred
A synthetic form of ACTH 1-24; increases plasma concentration of hydrocortisone in mares more rapidly and consistently than natural ACTH 1-24
Catalog No. 50-299-4393 Supplier Cayman Chemical Supplier No. 2391210 MG
A synthetic form of ACTH 1-24; increases plasma concentration of hydrocortisone in mares more rapidly and consistently than natural ACTH 1-24