Learn More
TARGETMOL CHEMICALS INC COT INHIBITOR-1 HYDROCHL 5MG
SDP
Catalog No. 502581635
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cot inhibitor-1 hydrochloride is an inhibitor of tumor progression loci-2 kinase (IC50 = 28 nM) and inhibits the production of TNF-(alpha) in human whole blood (IC50 = 5.7 nM).
Catalog No. 50-258-1635 Supplier TARGETMOL CHEMICALS INC Supplier No. T10865L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cot inhibitor-1 hydrochloride is an inhibitor of tumor progression loci-2 kinase (IC50 = 28 nM) and inhibits the production of TNF-(alpha) in human whole blood (IC50 = 5.7 nM).