Learn More
Medchemexpress LLC HY-32018 10mg Medchemexpress, Cot inhibitor-2 CAS:915363-56-3 Purity:>98%
SDP
Catalog No. 502029306
Encompass_Preferred
Medchemexpress, HY-32018 10mg Cot inhibitor-2 CAS:915363-56-3 Cot inhibitor-2 is a potent, selective and orally active cot (Tpl2/MAP3K8) inhibitor with an IC50 of 1.6 nM. Cot inhibitor-2 inhibts TNF-α production in LPS-stimulated human whole blood with an IC50 of 0.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9306 Supplier Medchemexpress LLC Supplier No. HY3201810MG
Medchemexpress, HY-32018 10mg Cot inhibitor-2 CAS:915363-56-3 Cot inhibitor-2 is a potent, selective and orally active cot (Tpl2/MAP3K8) inhibitor with an IC50 of 1.6 nM. Cot inhibitor-2 inhibts TNF-α production in LPS-stimulated human whole blood with an IC50 of 0.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.