Learn More
Carnabio Usa Inc COT(MAP3K8)/10UG/CATALYTIC
Catalog No. 507540741
Encompass
COT(MAP3K8), Catalytic domain, Wild type, Amino Acid 30-397, NP_005195.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0741 Supplier Carnabio Usa Inc Supplier No. 0730110
COT(MAP3K8), Catalytic domain, Wild type, Amino Acid 30-397, NP_005195.2, Expressed in Insect (sf21); 10microg