Learn More
Carnabio Usa Inc COT(MAP3K8)/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 073015
Encompass
COT(MAP3K8), Catalytic domain, Wild type, Amino Acid 30-397, NP_005195.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0386