Learn More
Sino Biological Recombinant Human MAP3K5/ASK1 Protein (GST Tag), Active
Catalog No. 502574382
Encompass_Preferred
Recombinant human ASK1(649-946) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Catalog No. 50-257-4382 Supplier Sino Biological Supplier No. M1311G500
Recombinant human ASK1(649-946) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.