Learn More
Medchemexpress LLC Cotadutide acetate | 99.3% | 3728.04 | 1 MG
SDP
Catalog No. 5000423124
Encompass_Preferred
Cotadutide acetate (MEDI0382 acetate) is a potent dual agonist of glucagon-like peptide-1 (GLP-1) and GCGR. It facilitates weight loss and glycemic control, and alleviates fibrosis. This product is for research use only and can be used in the study of obesity and type 2 diabetes (T2D).
  • Potent dual agonist for GLP-1 and GCGR
  • Facilitates weight loss and glycemic control
  • Alleviates fibrosis
  • Stimulates cAMP accumulation in β-cell lines and hepatocytes
  • Potentiates glucose-stimulated insulin secretion
  • Increases glucose output in hepatocytes
  • Induces mitochondrial turnover and enhances mitochondrial function
Catalog No. 50-004-23124 Supplier Medchemexpress LLC Supplier No. HYP2231A1MG
Cotadutide acetate (MEDI0382 acetate) is a potent dual agonist of glucagon-like peptide-1 (GLP-1) and GCGR. It facilitates weight loss and glycemic control, and alleviates fibrosis. This product is for research use only and can be used in the study of obesity and type 2 diabetes (T2D).
  • Potent dual agonist for GLP-1 and GCGR
  • Facilitates weight loss and glycemic control
  • Alleviates fibrosis
  • Stimulates cAMP accumulation in β-cell lines and hepatocytes
  • Potentiates glucose-stimulated insulin secretion
  • Increases glucose output in hepatocytes
  • Induces mitochondrial turnover and enhances mitochondrial function