Learn More
Selleck Chemical LLC COTI-2 5mg 1039455-84-9
SDP
Supplier:  Selleck Chemical LLC S85805MG
Encompass_Preferred
COTI-2 is an orally available third generation thiosemicarbazone and activator of mutant forms of the p53 protein, with potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8260