Learn More
Cayman Chemical -CotInIn-d3 10mg
SDP
Catalog No. 502971659
Encompass_Preferred
An internal standard for the quantification of cotinine by GC- or LC-MS
Catalog No. 50-297-1659 Supplier Cayman Chemical Supplier No. 1580510 MG
An internal standard for the quantification of cotinine by GC- or LC-MS