Learn More
Cayman Chemical (±)-Cotinine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 110952-70-0, 25 mg
SDP
Catalog No. 502972285
Encompass_Preferred
An internal standard for the quantification of cotinine by GC- or LC-MS
Catalog No. 50-297-2285 Supplier Cayman Chemical Supplier No. 1580525 MG
An internal standard for the quantification of cotinine by GC- or LC-MS