Learn More
Calbiotech Inc Cotinine ELISA Kit, solid phase competitive, 96 Tests (12x8 strip wells), 5-100 ng/ml standard range, sample volume 10 μl/well
Supplier:  Calbiotech Inc CO096D
Encompass_Preferred
Cotinine ELISA Kit, solid phase competitive, 96 Tests (12x8 strip wells), 5-100 ng/ml standard range, sample volume 10 μl/well
Catalog No. NC0365686