Learn More
Medchemexpress LLC Cotinine-d3 N-β-D-Glucuronide | 139427-57-9 | C16H17D3N2O7 | 1 MG
SDP
Catalog No. 5000374943
Encompass_Preferred
Cotinine-d3 N-β-D-Glucuronide is the deuterium-labeled form of Cotinine N-β-D-Glucuronide. It is a glucuronide conjugate of the tobacco metabolite Cotinine, promising for research related to tobacco carcinogen metabolism and cognitive function. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules are largely used as tracers for quantitation during drug development
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-74943 Supplier Medchemexpress LLC Supplier No. HY150159S1MG
Cotinine-d3 N-β-D-Glucuronide is the deuterium-labeled form of Cotinine N-β-D-Glucuronide. It is a glucuronide conjugate of the tobacco metabolite Cotinine, promising for research related to tobacco carcinogen metabolism and cognitive function. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules are largely used as tracers for quantitation during drug development
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs