Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cotinine N-β-D-glucuronide | 139427-57-9 | 96.6% | 352.34 | C16H20N2O7 | 1 MG
SDP

Catalog No. 5000285139
Encompass_Preferred

Cotinine N-β-D-glucuronide is the glucuronide conjugate of cotinine supplied as an analytical reference standard for metabolite and biomarker research related to nicotine exposure. The material is provided as a white to off-white solid with documented HPLC purity and elemental analysis suitable for analytical workflows.

  • Analytical reference standard for cotinine metabolite studies
  • HPLC purity 96.6%
  • White to off-white solid with documented appearance
  • CAS number 139427-57-9; formula C16H20N2O7; MW 352.34
  • Storage: sealed, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month)
  • COA includes HPLC and elemental analysis data for quality verification

Catalog No. 50-002-85139 Supplier Medchemexpress LLC Supplier No. HYW5881941MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cotinine N-β-D-glucuronide is the glucuronide conjugate of cotinine supplied as an analytical reference standard for metabolite and biomarker research related to nicotine exposure. The material is provided as a white to off-white solid with documented HPLC purity and elemental analysis suitable for analytical workflows.

  • Analytical reference standard for cotinine metabolite studies
  • HPLC purity 96.6%
  • White to off-white solid with documented appearance
  • CAS number 139427-57-9; formula C16H20N2O7; MW 352.34
  • Storage: sealed, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month)
  • COA includes HPLC and elemental analysis data for quality verification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.