Learn More
Cayman Chemical (−)-Cotinine, Purity: ≥98%, CAS: 486-56-6, 50 mg
SDP
Catalog No. 502971742
Encompass_Preferred
An alkaloid and active metabolite of (–)-nicotine; a weak nAChR agonist; reduces the number of NNK-induced dsDNA breaks in vitro; improves working memory and reduces immobility time in the forced swim test in chronically stressed mice; has been found as a contaminant in soils used to grow tobacco
Catalog No. 50-297-1742 Supplier Cayman Chemical Supplier No. 1531450 MG
An alkaloid and active metabolite of (–)-nicotine; a weak nAChR agonist; reduces the number of NNK-induced dsDNA breaks in vitro; improves working memory and reduces immobility time in the forced swim test in chronically stressed mice; has been found as a contaminant in soils used to grow tobacco