Learn More
Medchemexpress LLC Cotrimoxazole (trimethoprim/sulfamethoxazole 1:19) | 8064-90-2 | 99.8% | C24H29N7O6S | 1 ML
SDP
Catalog No. 5000353663
Encompass_Preferred
Cotrimoxazole (trimethoprim/sulfamethoxazole 1:19) is an antimicrobial agent, a mixture of trimethoprim and sulfamethoxazole with a 1:19 ratio. This product is for research use only.
  • Antimicrobial agent
  • Mixture of trimethoprim and sulfamethoxazole
  • 1:19 ratio of trimethoprim to sulfamethoxazole
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-53663 Supplier Medchemexpress LLC Supplier No. HY141619A10MM/1ML
Cotrimoxazole (trimethoprim/sulfamethoxazole 1:19) is an antimicrobial agent, a mixture of trimethoprim and sulfamethoxazole with a 1:19 ratio. This product is for research use only.
  • Antimicrobial agent
  • Mixture of trimethoprim and sulfamethoxazole
  • 1:19 ratio of trimethoprim to sulfamethoxazole
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month