Learn More
MSC Orion Cordage Tying Twine, Cotton, Plied Twist, 5/64 in., 880 ft., 1 ea.
Catalog No. 5000516595
Encompass
This spool twine is a high-quality, white cotton cord designed for various tying and bundling applications. Its durable plied twist construction offers reliable strength for general-purpose utility tasks in industrial and maintenance environments.
  • Constructed from 100% cotton material
  • Plied twist design for consistent performance
  • Breaking strength of 120 lb
  • Diameter of 5/64 in.
  • Supplied on a convenient spool for easy dispensing
  • Length of 880 ft. per pound
Catalog No. 50-005-16595 Supplier MSC Supplier No. 17233941
This spool twine is a high-quality, white cotton cord designed for various tying and bundling applications. Its durable plied twist construction offers reliable strength for general-purpose utility tasks in industrial and maintenance environments.
  • Constructed from 100% cotton material
  • Plied twist design for consistent performance
  • Breaking strength of 120 lb
  • Diameter of 5/64 in.
  • Supplied on a convenient spool for easy dispensing
  • Length of 880 ft. per pound