Learn More
Raybiotech Inc Sigmodon IFN-gamma ELISA Kit
Supplier:  Raybiotech Inc ELSIFNG1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.41 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Cotton rat (Sigmodon) IFN-gamma.
Catalog No. 50-195-0317