Learn More
Med Vet International Cotton Stockinette, J0291DC, 4in x 25 Yards, Each
Catalog No. 5000582251
Encompass_Preferred
Useful for protecting extremities prior to splinting. Use as non-adhesive liner under a splint for small and large animals. Stockinette is removable and washable.
Catalog No. 50-005-82251 Supplier Med Vet International Supplier No. JOR291DC
Useful for protecting extremities prior to splinting. Use as non-adhesive liner under a splint for small and large animals. Stockinette is removable and washable.