Learn More
Electron Microscopy Sciences Cotton Tip Applicator, Anti Static, Lint Free
SDP
Catalog No. 50949315
Encompass_Preferred
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft. The pointed head size is 3/16"(d) tapering to 1/32"(d) x 5/8 "(L). Regular head size 3/16(d) x 5/8 "(L). Packed 100 sticks in a sealed bag. Anti-Static, Lint-Free. Regular Tip 15⁄64" Dia.x 11⁄16"L. Polypropylene Shaft.
Catalog No. 50-949-315 Supplier Electron Microscopy Sciences Supplier No. 72306
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft. The pointed head size is 3/16"(d) tapering to 1/32"(d) x 5/8 "(L). Regular head size 3/16(d) x 5/8 "(L). Packed 100 sticks in a sealed bag. Anti-Static, Lint-Free. Regular Tip 15⁄64" Dia.x 11⁄16"L. Polypropylene Shaft.