Learn More
Andwin Scientific COTTON TIP APPLICATOR 1000/PKG
SDP
Catalog No. NC9497626
Encompass_Preferred
The Cotton-Tipped Wooden Applicators are efficient product for use in bacteriology labs for inoculating broth and plated media, and obtaining culture specimens. The applicators can also be utilized for sanitizing delicate laboratory equipments or instruments. The wooden handle measures 152.4 mm (6.00") x 2.5 mm (0.100") while the head measures 15.9 mm (0.625") x 4.7 mm (0.188"). This item is sold in a pack of 1000.
This item is not returnable. View return policy
Catalog No. NC9497626 Supplier Andwin Scientific Supplier No. 10806000
The Cotton-Tipped Wooden Applicators are efficient product for use in bacteriology labs for inoculating broth and plated media, and obtaining culture specimens. The applicators can also be utilized for sanitizing delicate laboratory equipments or instruments. The wooden handle measures 152.4 mm (6.00") x 2.5 mm (0.100") while the head measures 15.9 mm (0.625") x 4.7 mm (0.188"). This item is sold in a pack of 1000.